Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NOBOX Antibody (OTI4C5), Janelia Fluor™ 646, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273028JF646
Description
NOBOX Monoclonal specifically detects NOBOX in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
NOBOX | |
Monoclonal | |
Janelia Fluor 646 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
NOBOX | |
Recombinant protein fragment corresponding to amino acids 1-271 of human NOBOX (NP_001073882) produced in E.coli. | |
0.1 mL | |
135935.0 | |
Store at 4°C in the dark. |
Western Blot, Immunohistochemistry | |
OTI4C5 | |
Western Blot, Immunohistochemistry | |
NOBOX NOBOX oogenesis homeobox, OG2, OG-2, OG2X, POF5, TCAG_12042 | |
Mouse | |
Affinity Purified | |
Primary | |
Human | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction