Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NOBOX Monoclonal Antibody (OTI4C5)

Mouse Monoclonal Antibody

Supplier:  Invitrogen™ MA526979

Catalog No. PIMA526979


Only null left
Add to Cart

Description

Description

NOBOX Monoclonal Antibody for Western Blot, IHC (P)

NOBOX is a transcription factor which may play a role in oogenesis. It binds preferentially to the DNA sequences 5'-TAATTG-3', 5'-TAGTTG-3' and 5'-TAATTA-3'.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

NOBOX
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
O60393
NOBOX
Human recombinant protein fragment corresponding to amino acids 1-271 of NOBOX produced in E.coli.
100 μL
Primary
Human
Antibody
IgG1
Immunohistochemistry (Paraffin), Western Blot
OTI4C5
Unconjugated
NOBOX
Homeobox protein NOBOX; homeodomain-containing protein OG-2; LOW QUALITY PROTEIN: homeobox protein NOBOX; newborn ovary homeobox protein; newborn ovary homeobox-encoding; Nobox; NOBOX oogenesis homeobox; OG2; OG-2; OG2 homeobox; Og2x; Oocyte-specific homeobox protein; POF5; TCAG_12042
Mouse
Affinity chromatography
RUO
135935
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.