Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ NOBOX Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA568881
Description
This target displays homology in the following species: Mouse: 100%.
NOBOX is a transcription factor which may play a role in oogenesis. It binds preferentially to the DNA sequences 5'-TAATTG-3', 5'-TAGTTG-3' and 5'-TAATTA-3'.
Specifications
NOBOX | |
Polyclonal | |
Unconjugated | |
NOBOX | |
Homeobox protein NOBOX; homeodomain-containing protein OG-2; LOW QUALITY PROTEIN: homeobox protein NOBOX; newborn ovary homeobox protein; newborn ovary homeobox-encoding; Nobox; NOBOX oogenesis homeobox; OG2; OG-2; OG2 homeobox; Og2x; Oocyte-specific homeobox protein; POF5; TCAG_12042 | |
Rabbit | |
Affinity chromatography | |
RUO | |
18291 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q8VIH1 | |
NOBOX | |
synthetic peptide corresponding to a region of mouse NOBOX (aa 201-250). | |
100 μL | |
Primary | |
Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction