Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NOBOX Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA541700
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA541700 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA541700 Supplier Thermo Scientific Supplier No. PA541700
Add to Cart
Add to Cart

NOBOX Polyclonal Antibody

NOBOX Polyclonal Antibody for Western Blot, IHC

TRUSTED_SUSTAINABILITY

Specifications

Antigen NOBOX
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene NOBOX
Gene Accession No. O60393
Gene Alias Homeobox protein NOBOX; homeodomain-containing protein OG-2; LOW QUALITY PROTEIN: homeobox protein NOBOX; newborn ovary homeobox protein; newborn ovary homeobox-encoding; Nobox; NOBOX oogenesis homeobox; OG2; OG-2; OG2 homeobox; Og2x; Oocyte-specific homeobox protein; POF5; TCAG_12042
Gene Symbols NOBOX
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human NOBOX (aa 37-86).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 135935
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.