Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NOC2L Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA550467
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA550467 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA550467 Supplier Invitrogen™ Supplier No. PA550467
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The antibody detects endogenous levels of total INHAT-2 / NOC2L protein.

INHAT (inhibitor of acetyltransferases) complex binds to histones and masks them from being substrate for acetyltransferase. Endogenous INHAT subunits, which include the Set/TAF-Ibeta oncoprotein, associate with chromatin in vivo and can block coactivator mediated transcription when transfected in cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NOC2L
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% sodium azide; pH 7.4
Gene NOC2L
Gene Accession No. Q9Y3T9
Gene Alias AA410003; AF155546; DKFZp564C186; FLJ35172; hypothetical protein LOC508638; NET15; NET7; NIR; NOC2 like nucleolar associated transcriptional repressor; NOC2L; NOC2L nucleolar associated transcriptional repressor; NOC2-like; NOC2-like nucleolar associated transcriptional repressor; NOC2-like protein; novel INHAT (inhibitor of histone acetyltransferase) repressor; novel INHAT repressor; nucleolar complex associated 2 homolog; nucleolar complex associated 2 homolog (S. cerevisiae); nucleolar complex protein 2 homolog; PPP1R112; Protein NOC2 homolog; protein phosphatase 1, regulatory subunit 12; RGD1309387
Gene Symbols NOC2L
Host Species Rabbit
Immunogen Fusion protein corresponding to a region derived from internal residues of human nucleolar complex associated 2 homolog.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26155
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.