Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NOC3L Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA536501
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA536501 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA536501 Supplier Invitrogen™ Supplier No. PA536501
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This antibody detects endogenous protein at a molecular weight of 93 kDa. Purity is >95% by SDS-PAGE.

GADD 153, a growth arrest and DNA damage-inducible gene, encodes a C/EBP-related nuclear protein. This protein has also been designated C/EBP-homologous protein (CHOP-10 or C/EBP zeta). GADD 153 expression is induced by a variety of cellular stresses, inducing nutrient deprivation and metabolic perturbations. GADD 153 functions to block cells in G1 to S phase during cell cycle progression and acts by dimerizing with other C/EBP proteins to direct GADD 153 dimers away from classical C/EBP binding sites, recognizing instead unique nonclassical sites. Thus, GADD 153 acts as a negative modulator of C/EBP-like proteins in certain terminally differentiated cells. GADD 153 belongs to the CBF/MAK21 family, which also includes NOC2L, NOC3L and NOC4L. NOC3L, also designated factor for adipocyte differentiation 24 or Fad24, promotes adipogenesis by controlling DNA replication during the early stages of mitotic clonal expansion (MCE).
TRUSTED_SUSTAINABILITY

Specifications

Antigen NOC3L
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene NOC3L
Gene Accession No. Q8VI84, Q8WTT2
Gene Alias AD24; AF233884; C10orf117; factor for adipocyte differentiation 24; Fad24; NOC3 like DNA replication regulator; NOC3 protein homolog; NOC3L; NOC3L DNA replication regulator; NOC3-like DNA replication regulator; NOC3-like protein; nucleolar complex associated 3 homolog; nucleolar complex associated 3 homolog (S. cerevisiae); nucleolar complex protein 3 homolog; Nucleolar complex-associated protein 3-like protein; RGD1560656
Gene Symbols NOC3L
Host Species Rabbit
Immunogen Synthetic peptide corresponding to amino acids 417-436 of human NOC3L.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 361753, 57753, 64318
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.