Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NOC4L Antibody (PCRP-NOC4L-1E3), mFluor Violet 500 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP308308MFV500
Description
NOC4L Monoclonal antibody specifically detects NOC4L in Human samples. It is validated for Flow CytometrySpecifications
NOC4L | |
Monoclonal | |
mFluor Violet 500 SE | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
79050 | |
Store at 4C in the dark. | |
IgG1 |
Flow Cytometry | |
PCRP-NOC4L-1E3 | |
Flow Cytometry | |
MGC3162, NET49, NOC4, NOC4 protein homolog, NOC4-like protein, nucleolar complex associated 4 homolog (S. cerevisiae), nucleolar complex protein 4 homolog, Nucleolar complex-associated protein 4-like protein | |
Recombinant full-length human NOC4L protein (Uniprot: Q9BVI4) | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction