Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NOC4L Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565787
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565787 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565787 Supplier Invitrogen™ Supplier No. PA565787
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GVRRALGRRLE AVLASRSEAN AVFDILAVLQ SEDQEEIQEA VRTCSRLFGA LLERGELFVG QLPSEEMVMT GSQGATRKYK VWMRHRYHSC CN Highest antigen sequence identity to the following orthologs - mouse 76%, rat 78%.

NOC4L plays a specific role in biogenesis of 40 S subunit of ribosome.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NOC4L
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene NOC4L
Gene Accession No. Q9BVI4
Gene Alias AI326906; NET49; NOC4; NOC4 protein homolog; NOC4L; NOC4-like protein; nucleolar complex associated 4 homolog; nucleolar complex associated 4 homolog (S. cerevisiae); nucleolar complex protein 4 homolog; Nucleolar complex-associated protein 4-like protein; RGD1310661; UTP19
Gene Symbols NOC4L
Host Species Rabbit
Immunogen Recombinant Human NOC4L. Recombinant protein control fragment (Product #RP-104965).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79050
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.