Learn More
Invitrogen™ NOC4L Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5110037
Description
Immunogen sequence: MEREPGAAGV RRALGRRLEA VLASRSEANA VFDILAVLQS EDQEEIQEAV RTCSRLFGAL LERGELFVGQ LPSEEMVMTG SQGATRKYKV WMRHRYHSCC NRLGELLGHP SFQVKELALS ALLKFVQLEG AHPLEKSKWE GNYLFPRELF KLVVGGLLSP EEDQSLLLSQ FREYLDYDDT RYHTMQAAVD AVARVTGQHP EVPPAFWNNA FTLLSAVSLP RREPTVSSFY VKRAELWDTW KVAHLKEHRR VFQAMWLSFL.
NOC4L plays a specific role in biogenesis of 40 S subunit of ribosome.
Specifications
NOC4L | |
Polyclonal | |
Unconjugated | |
NOC4L | |
AI326906; NET49; NOC4; NOC4 protein homolog; NOC4L; NOC4-like protein; nucleolar complex associated 4 homolog; nucleolar complex associated 4 homolog (S. cerevisiae); nucleolar complex protein 4 homolog; Nucleolar complex-associated protein 4-like protein; RGD1310661; UTP19 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
79050 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
1.26 mg/mL | |
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
Q9BVI4 | |
NOC4L | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-260 of human NOC4L (NP_076983.1). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.