Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nocturnin Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501981370
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-1370 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-1370 Supplier Bioss Supplier No. BS11444R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CCRN4L is highly similar to Nocturnin, identified as a circadian clock regulated gene in Xenopus laevis. This protein and Nocturnin protein share similarity with the C-terminal domain of a yeast transcription factor, carbon catabolite repression 4 (CCR4).

Specifications

Antigen Nocturnin
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene NOCT
Gene Accession No. Q9UK39
Gene Alias AU043840; carbon catabolite epression 4 homolog; carbon catabolite repression 4 homolog; Carbon catabolite repression 4-like protein; Ccr4; CCR4 carbon catabolite repression 4-like; CCR4 carbon catabolite repression 4-like (S. cerevisiae); CCR4 carbon catabolite repression 4-like B; CCR4 protein homolog; Ccr4c; CCR4L; CCR4-like (carbon catabolite repression 4, S.cerevisiae); Ccrn4l; Ccrn4lb; circadian deadenylase NOC; MGC142054; MGC142060; MGC4120817; MGC78549; Noc; NOCT; nocturin; Nocturnin
Gene Symbols NOCT
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human Nocturnin.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25819
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.