Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NOD1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 3 publications
$181.90 - $402.00
Specifications
| Antigen | NOD1 |
|---|---|
| Dilution | Western Blot 1:1000-1:2500, Immunohistochemistry-Paraffin 1:100 |
| Applications | Western Blot, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB10056878T
![]() |
Novus Biologicals
NB10056878SS |
0.025 mL |
Each for $181.90
|
|
|||||
NB10056878
![]() |
Novus Biologicals
NB10056878 |
0.1 mL |
Each for $402.00
|
|
|||||
Description
NOD1 Polyclonal specifically detects NOD1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| NOD1 | |
| Western Blot, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Apoptosis, Asthma, Autophagy, Immunology | |
| Whole antisera with 50% Glycerol with 0.025% Sodium Azide | |
| CARD4NLR family, CARD domain containing 1, caspase recruitment domain family, member 4, Caspase recruitment domain-containing protein 4, CLR7.1, NLRC1, nucleotide-binding oligomerization domain containing 1, nucleotide-binding oligomerization domain, leucine rich repeat and CARD domaincontaining 1, nucleotide-binding oligomerization domain-containing protein 1 | |
| NOD1 | |
| IgG | |
| Unpurified | |
| Cross reacts with Human. Predicted to cross react with New World Monkey and Mouse. |
| Western Blot 1:1000-1:2500, Immunohistochemistry-Paraffin 1:100 | |
| Polyclonal | |
| Antisera | |
| RUO | |
| Human | |
| Q9Y239 | |
| 10392 | |
| Amino acids 930-949 of human NOD1 protein were used as the immunogen for this antibody. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title