Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NOD1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. 01676643
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-676-643 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-676-643 Supplier Invitrogen™ Supplier No. PA141036
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Suggested positive control: antigen standard for NOD1 (transient overexpression lysate), recombinant fusion protein. Predicted to cross react with New World Monkey and Mouse.

NOD1 is a member of the NOD (nucleotide-binding oligomerization domain) family, a group of proteins that are involved in innate immune defense. NOD1 contains an N-terminal caspase recruitment domain (CARD), a centrally located nucleotide-binding domain (NBD), and ten tandem leucine-rich repeats (LRRs) in its C-terminus. The CARD is involved in apoptotic signaling, and NOD1 activates caspase-9 and NF-kappa-B. LRRs participate in protein-protein interactions, and mutations in the NBD may affect the process of oligomerization and subsequent function of the LRR domain. This protein is an intracellular pattern-recognition receptor (PRR) that initiates inflammation in response to a subset of bacteria through the detection of bacterial diaminopimelic acid.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NOD1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation Whole serum with 50% glycerol and 0.025% sodium azide
Gene NOD1
Gene Accession No. Q8BHB0, Q9Y239
Gene Alias C230079P11; Card4; caspase recruitment domain 4; caspase recruitment domain family, member 4; caspase recruitment domain-containing protein 4; CLR7.1; F830007N14Rik; NLR family, CARD domain containing 1; Nlrc1; Nod1; nucleotide binding oligomerization domain containing 1; nucleotide-binding oligomerization domain containing 1; nucleotide-binding oligomerization domain, leucine rich repeat and CARD domain containing 1; nucleotide-binding oligomerization domain-containing protein 1; RGD1562269
Gene Symbols NOD1
Host Species Rabbit
Immunogen Amino acids 930-949 of human NOD1 protein.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10392, 107607
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.