Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NOD1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA521015
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA521015 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA521015 Supplier Invitrogen™ Supplier No. PA521015
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

A suggested positive control is EL4 cell lysate. PA5-21015 can be used with blocking peptide PEP-1129.

NOD1 is a member of the NOD (nucleotide-binding oligomerization domain) family, a group of proteins that are involved in innate immune defense. NOD1 contains an N-terminal caspase recruitment domain (CARD), a centrally located nucleotide-binding domain (NBD), and ten tandem leucine-rich repeats (LRRs) in its C-terminus. The CARD is involved in apoptotic signaling, and NOD1 activates caspase-9 and NF-kappa-B. LRRs participate in protein-protein interactions, and mutations in the NBD may affect the process of oligomerization and subsequent function of the LRR domain. This protein is an intracellular pattern-recognition receptor (PRR) that initiates inflammation in response to a subset of bacteria through the detection of bacterial diaminopimelic acid.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NOD1
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene NOD1
Gene Accession No. Q8BHB0, Q9Y239
Gene Alias C230079P11; Card4; caspase recruitment domain 4; caspase recruitment domain family, member 4; caspase recruitment domain-containing protein 4; CLR7.1; F830007N14Rik; NLR family, CARD domain containing 1; Nlrc1; Nod1; nucleotide binding oligomerization domain containing 1; nucleotide-binding oligomerization domain containing 1; nucleotide-binding oligomerization domain, leucine rich repeat and CARD domain containing 1; nucleotide-binding oligomerization domain-containing protein 1; RGD1562269
Gene Symbols NOD1
Host Species Rabbit
Immunogen A 16 amino acid peptide near the carboxy end of human NOD1.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10392, 107607, 500133
Target Species Human, Mouse, Rat
Content And Storage Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.