Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ NOD1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA521981
Description
Recommended positive controls: 293T, A431, H1299, HeLa, HepG2, Raji. Predicted reactivity: Mouse (81%), Rat (82%), Pig (87%), Bovine (87%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
NOD1 is a member of the NOD (nucleotide-binding oligomerization domain) family, a group of proteins that are involved in innate immune defense. NOD1 contains an N-terminal caspase recruitment domain (CARD), a centrally located nucleotide-binding domain (NBD), and ten tandem leucine-rich repeats (LRRs) in its C-terminus. The CARD is involved in apoptotic signaling, and NOD1 activates caspase-9 and NF-kappa-B. LRRs participate in protein-protein interactions, and mutations in the NBD may affect the process of oligomerization and subsequent function of the LRR domain. This protein is an intracellular pattern-recognition receptor (PRR) that initiates inflammation in response to a subset of bacteria through the detection of bacterial diaminopimelic acid.
Specifications
NOD1 | |
Polyclonal | |
Unconjugated | |
NOD1 | |
C230079P11; Card4; caspase recruitment domain 4; caspase recruitment domain family, member 4; caspase recruitment domain-containing protein 4; CLR7.1; F830007N14Rik; NLR family, CARD domain containing 1; Nlrc1; Nod1; nucleotide binding oligomerization domain containing 1; nucleotide-binding oligomerization domain containing 1; nucleotide-binding oligomerization domain, leucine rich repeat and CARD domain containing 1; nucleotide-binding oligomerization domain-containing protein 1; RGD1562269 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10392 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
Q9Y239 | |
NOD1 | |
Recombinant protein encompassing a sequence within the C-terminus region of human NOD1. The exact sequence is proprietary. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction