Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NOD1 Sheep anti-Human, Polyclonal, R&D Systems™
Sheep Polyclonal Antibody
Specifications
Antigen | NOD1 |
---|---|
Dilution | Immunohistochemistry 1-15 ug/mL |
Applications | Immunohistochemistry |
Classification | Polyclonal |
Conjugate | Unconjugated |
Specifications
NOD1 | |
Immunohistochemistry | |
Unconjugated | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
CARD4, CARD4NLR family, CARD domain containing 1, caspase recruitment domain family, member 4, Caspase recruitment domain-containing protein 4, CLR7.1, NLRC1, nucleotide-binding oligomerization domain containing 1, nucleotide-binding oligomerization domain, leucine rich repeat and CARD domaincontaining 1, nucleotide-binding oligomerization domain-containing protein 1 | |
E. coli-derived recombinant human NOD1, Lys830-Phe953, Accession # Q9Y239 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects human NOD1. |
Immunohistochemistry 1-15 ug/mL | |
Polyclonal | |
Sheep | |
Human | |
Q9Y239 | |
10392 | |
IgG | |
Antigen Affinity-purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title