Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NOD2 Mouse anti-Human, Mouse, Clone: 2D9, Abnova™

Catalog No. 89934703 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-934-703 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-934-703 Supplier Abnova Corporation Supplier No. MAB2303
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against partial recombinant NOD2.

This gene is a member of the Nod1/Apaf-1 family and encodes a protein with two caspase recruitment (CARD) domains and six leucine-rich repeats (LRRs). The protein is primarily expressed in the peripheral blood leukocytes. It plays a role in the immune response to intracellular bacterial lipopolysaccharides (LPS) by recognizing the muramyl dipeptide (MDP) derived from them and activating the NFKB protein. Mutations in this gene have been associated with Crohn disease and Blau syndrome. [provided by RefSeq]

Specifications

Antigen NOD2
Applications Western Blot
Classification Monoclonal
Clone 2D9
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against partial recombinant NOD2.
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS (0.02% sodium azide)
Gene Alias ACUG/BLAU/CARD15/CD/CLR16.3/IBD1/NLRC2/NOD2B/PSORAS1
Gene Symbols NOD2
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acids 28-301 of human NOD2.
Purification Method Protein G purification
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64127
Test Specificity This antibody is specific to NOD2 protein.
Target Species Human, Mouse
Content And Storage Store at 4°C for short term. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.