Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NOD2 Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA523101

Catalog No. PIPA523101


Only null left
Add to Cart

Description

Description

The amino acid sequence used as immunogen is 95% homologous in Lar gibbon (Hylobates lar), 97% in chimpanzee.

NOD2 is a member of the apoptosis regulating protein family that includes caspase recruitment-domains, as well as Apaf-1 and NOD1. It contains two N-terminal CARDs, a nucleotide binding domain (NBD), and multiple C-terminal leucine-rich repeats (LRRs). NOD2 is expressed in monocytes (whereas NOD1 is expressed in multiple tissues). NOD2 plays a role in regulating NF-kappaB, acts as an intracellular receptor for bacterial lipopolysaccharides, and contributes to inflammatory bowel disease (IBD) and Crohn's disease.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

NOD2
Polyclonal
Unconjugated
NOD2
ACUG; BLAU; CARD15; caspase recruitment domain family, member 15; caspase recruitment domain protein 15; caspase recruitment domain-containing protein 15; CD; CLR16.3; F830032C23Rik; IBD1; Inflammatory bowel disease protein 1; NLR family, CARD domain containing 2; Nlrc2; NOD 2; NOD2; NOD-2; NOD2 protein; NOD2B; NOD-like receptor C2; nucleotide binding oligomerization domain containing 2; nucleotide-binding oligomerization domain 2; nucleotide-binding oligomerization domain containing 2; nucleotide-binding oligomerization domain, leucine rich repeat and CARD domain containing 2; nucleotide-binding oligomerization domain-containing protein 2; PSORAS1
Rabbit
100 μL
Primary
Human, Mouse
Antibody
IgG
Immunoprecipitation, Western Blot
Conc. Not Determined
Whole serum with 50% glycerol and 0.05% sodium azide
Q8K3Z0, Q9HC29
NOD2
Synthetic peptide corresponding to amino acids 125-162 of human Nod2.
RUO
257632, 64127
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

WARNING: Cancer - www.P65Warnings.ca.gov
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.