Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NOD2 Rat anti-Mouse, Clone: 26mNOD2, Abnova™

Catalog No. 89934702 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-934-702 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-934-702 Supplier Abnova Corporation Supplier No. MAB12647

Rat monoclonal antibody raised against mouse Nod2.

Specifications

Antigen NOD2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 26mNOD2
Conjugate Unconjugated
Formulation In PBS, pH 7.2 (0.09% sodium azide).
Gene Alias ACUG, BLAU, CD, Card15, F830032C23Rik, IBD1, Nlrc2
Gene Symbols Nod2
Host Species Rat
Immunogen Mouse Nod2
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 257632
Target Species Mouse
Content And Storage Store at 4°C. Aliquot to avoid repeated freezing and thawing.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.