Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ NOD3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5109202
Description
Positive Control: Human thymus tissue lysates, HUVEC, MCF-7, SH-SY5Y, human tonsil tissue, human colon cancer tissue, human spleen tissue, mouse colon tissue.
NOD3 is a member of the NOD (nucleotide-binding oligomerization domain) family, a group of proteins that are involved in innate immune defense. NOD3 also known as NLR family CARD containing 3 (NLRC3), is predominantly expressed in the immune system, particularly in T lymphocytes, and its expression is strongly down-regulated following stimulation of the T-cell receptor complex and CD28, suggesting that NOD3 plays a role in attenuating the activation of T cells. NOD3 inhibits NF-βB, AP-1 and NFAT transcriptional activation in Jurkat T cells downstream of CD3/CD28 stimulation or treatment with PMA/ionomycin and decreases IL-2 and CD25 mRNA induction in activated cells.
Specifications
NOD3 | |
Polyclonal | |
Unconjugated | |
NLRC3 | |
CARD15-like protein; caterpillar 16.2; Caterpiller 16.2; caterpiller protein 16.2; CLR16.2; D230007K08Rik; mFLJ00348; NACHT, LRR and CARD domains-containing protein 3; NLR family CARD domain containing 3; NLR family CARD domain-containing protein 3; NLR family, CARD domain containing 3; NLR family, CARD domain containing 3, pseudogene 1; Nlrc3; Nlrc3-ps1; NOD3; NOD-like receptor C3; Nucleotide-binding oligomerization domain protein 3; nucleotide-binding oligomerization domain, leucine rich repeat and CARD domain containing 3; protein NLRC3; RGD1304902 | |
Rabbit | |
Affinity chromatography | |
RUO | |
197358 | |
-20°C, Avoid Freeze/Thaw Cycles, store in dark | |
Liquid |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4 | |
Q7RTR2 | |
NLRC3 | |
Synthetic peptide within Human NLRC3 aa 1-50. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction