Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NOD4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566236
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566236 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566236 Supplier Invitrogen™ Supplier No. PA566236
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GLVRCFSTLQW LFRLDISFES QHILLRGDKT SRDMWATGSL PDFPAAAKFL GFRQRCIPRS LCLSECPLEP PSLTRLCATL KDCPGPLELQ LSCEFLSDQ Highest antigen sequence identity to the following orthologs - mouse 55%, rat 50%.

NOD4 is a member of the NOD (nucleotide-binding oligomerization domain) family, a group of proteins that are involved in innate immune defense. NOD4 contains a CARD-like domain, a central NOD domain and a large LRR region. NOD4, an IFN-gamma-inducible nuclear protein, plays a role in homeostatic control of innate immunity and in antiviral defense mechanisms. As a key negative regulator of NF-kappa-B and type I interferon signaling, NOD4 may be a useful target for manipulating immune responses against infectious or inflammation-associated diseases, including cancer.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NOD4
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene NLRC5
Gene Accession No. Q86WI3
Gene Alias AI451557; AK220210; Caterpiller protein 16.1; CLR16.1; copine II; Cpne2; NLR family CARD domain containing 5; NLR family, CARD domain containing 5; Nlrc5; NOD27; NOD4; NOD-like receptor C5; Nucleotide-binding oligomerization domain protein 27; Nucleotide-binding oligomerization domain protein 4; nucleotide-binding oligomerization domain, leucine rich repeat and CARD domain containing 5; nucleotide-binding oligomerization domains 27; protein NLRC5
Gene Symbols NLRC5
Host Species Rabbit
Immunogen Recombinant Human NOD4. Recombinant protein control fragment (Product #RP-106918).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84166
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.