Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Nodal Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA597289
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA597289 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA597289 Supplier Invitrogen™ Supplier No. PA597289
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SNLSQEQRQL GGSTLLWEAE SSWRAQEGQL SWEWGKRHRR HHLPDRSQLC RKVKFQVDFN LIGWGSWIIY PKQYNAYRCE GECPNPVGEE FHPTNHAYIQ SLLKRYQPHR VPSTCCAPVK TKPLSMLYVD NGRVLLDHHK DMIVEECGCL; Positive Samples: SH-SY5Y, HepG2, 293T, Rat liver.

NODAL is an important TGF-beta family protein in cellular differentiation. While constitutively expressed in many developing tissues, NODAL is problematically associated with the differentiation and development of many cancers. The expression of NODAL is regulated by a Notch signaling pathway, which when disrupted contributes to the aggressive behavior of metastatic cancer cells, particularly in melanoma. The abundance of NODAL expression in melanoma can help determine the stage of cancer and metastatic type/progression when tested in IHC. NODAL interactions with Cripto also predispose toward tumor progression. Overexpression of NODAL is also associated with preeclamptic placenta.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Nodal
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.54 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene NODAL
Gene Accession No. Q96S42
Gene Alias early embryo mesoderm formation; HTX5; MGC138230; Nodal; nodal growth differentiation factor; nodal homolog; Tg.413d; transgenic lethal 413.d
Gene Symbols NODAL
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 198-347 of human NODAL (NP_0605253).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 294503, 4838
Target Species Human, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.