Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Nodal Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5111214
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5111214 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5111214 Supplier Invitrogen™ Supplier No. PA5111214
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PTEGSLAIEI FHQPKPDTEQ ASDSCLERFQ MDLFTVTLSQ VTFSLGSMVL EVTRPLSKWL KHPGALEKQM SRV Highest antigen sequence indentity to the following orthologs: Mouse - 64%, Rat - 64%.

NODAL is an important TGF-beta family protein in cellular differentiation. While constitutively expressed in many developing tissues, NODAL is problematically associated with the differentiation and development of many cancers. The expression of NODAL is regulated by a Notch signaling pathway, which when disrupted contributes to the aggressive behavior of metastatic cancer cells, particularly in melanoma. The abundance of NODAL expression in melanoma can help determine the stage of cancer and metastatic type/progression when tested in IHC. NODAL interactions with Cripto also predispose toward tumor progression. Overexpression of NODAL is also associated with preeclamptic placenta.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Nodal
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene NODAL
Gene Accession No. Q96S42
Gene Alias early embryo mesoderm formation; HTX5; MGC138230; Nodal; nodal growth differentiation factor; nodal homolog; Tg.413d; transgenic lethal 413.d
Gene Symbols NODAL
Host Species Rabbit
Immunogen Recombinant Protein corresonding to Human Nodal. Recombinant protein control fragment (Product #RP-91947).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4838
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.