Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Nodal Rabbit anti-Human, Rat, Clone: S06-5A3, Novus Biologicals™

Rabbit Monoclonal Antibody
$263.00 - $573.00
Specifications
| Antigen | Nodal |
|---|---|
| Clone | S06-5A3 |
| Dilution | Western Blot 1:1000-1:5000 |
| Applications | Western Blot |
| Classification | Monoclonal |
Description
Nodal Monoclonal antibody specifically detects Nodal in Human, Rat samples. It is validated for Western BlotSpecifications
| Nodal | |
| Western Blot 1:1000-1:5000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Rat | |
| MGC138230, nodal homolog, nodal homolog (mouse), nodal, mouse, homolog | |
| A synthetic peptide of human Nodal (Uniprot#: Q96S42) | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| S06-5A3 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Cytokine Research, Stem Cell Signaling Pathway | |
| 50nM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA with 0.01% Sodium Azide | |
| 4838 | |
| IgG | |
| Affinity-purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title