Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Nofetumomab Recombinant Mouse Monoclonal Antibody
GREENER_CHOICE

Catalog No. PIMA541755
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA541755 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA541755 Supplier Invitrogen™ Supplier No. MA541755
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Recombinant Monoclonal Antibody

Endotoxin level < 0.01EU/μg by LAL method Use a manual defrost freezer and avoid repeated freeze thaw cycles. Store at 2 to 8 °C for one week. Store at -20 to -80 °C for twelve months from the date of receipt.

Technetium Tc-99m nofetumomab merpentan (Tc-99m nm) consists of a Fab fragment of an IgG2b of the pancarcinoma murine antibody NR-LU-10. 1The NR-LU-10 antibody is directed against a 40 kDa glycoprotein antigen expressed in a variety of cancers and some normal tissues. LabelTc-99m nm was developed by Boehringer Ingelheim Pharma KG and FDA approved on September 14, 1992. It was after discontinued on August 13, 2013, but in the 2018 FDA submission list, it can be found as a substance type II (pharmaceutical substance) with an active status. The mechanism of action in Tc-99m nm is ruled by the presence of nofetumomab, which can recognize the pancarcinoma antigen EpCAM and/or CD20/MS4A1, 2and merpentan, that acts as a linker for the binding of technetium.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Nofetumomab
Applications ELISA, SDS-Page
Classification Recombinant Monoclonal
Concentration 2.2 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene Alias Verluma; Technetium (99mTc) nofetumomab merpentan
Host Species Mouse
Immunogen Human CD20/MS4A1.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2b F(ab) κ
Target Molecule Chemical
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.