Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Noggin Antibody (OTI1H8), Alexa Fluor™ 594, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273030AF594
Description
Noggin Monoclonal specifically detects Noggin in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
Noggin | |
Monoclonal | |
Alexa Fluor 594 | |
50 mM sodium borate with 0.05% sodium azide | |
NOG | |
Recombinant protein fragment corresponding to amino acids 28-232 of human Noggin (NP_005441) produced in E.coli. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG2b |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
OTI1H8 | |
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
noggin, SYM1, symphalangism 1 (proximal), synostoses (multiple) syndrome 1, SYNS1 | |
Mouse | |
Affinity Purified | |
Biologically Active Proteins, Neuroscience, Stem Cell Markers, Stem Cell Signaling Pathway | |
9241.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction