Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Noggin Antibody (OTI1H8) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273030
Description
Noggin Monoclonal specifically detects Noggin in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
| Noggin | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1:20000-40000, Immunohistochemistry 1:50, Immunocytochemistry/Immunofluorescence 1:100 | |
| noggin, SYM1, symphalangism 1 (proximal), synostoses (multiple) syndrome 1, SYNS1 | |
| Mouse | |
| 23.7 kDa | |
| 100 μg | |
| Primary | |
| Reconstitute with PBS (pH 7.3). To use this carrier-free antibody for conjugation experiments, another round of desalting is highly recommended. | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG2b |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| OTI1H8 | |
| Unconjugated | |
| Lyophilized from PBS (pH 7.3) with 8% Trehalose with No Preservative | |
| NOG | |
| Human recombinant protein fragment corresponding to amino acids 28-232 of human noggin (NP_005441) produced in E.coli. | |
| Affinity Purified | |
| Biologically Active Proteins, Neuroscience, Stem Cell Markers, Stem Cell Signaling Pathway | |
| 9241 | |
| Human, Mouse, Rat | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction