Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Noggin Antibody (OTI1H8), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP147881
Description
Noggin Monoclonal specifically detects Noggin in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
Noggin | |
Monoclonal | |
1 mg/mL | |
Western Blot 1:20000-40000, Immunohistochemistry 1:50, Immunocytochemistry/Immunofluorescence 1:100, Immunohistochemistry-Paraffin 1:50 | |
NOG | |
Human recombinant protein fragment corresponding to amino acids 28-232 of human noggin (NP_005441) produced in E.coli. | |
Affinity Purified | |
RUO | |
Primary | |
This antibody is specific for Homo sapiens noggin (NOG). | |
Store at -20°C. Avoid freeze-thaw cycles. |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
OTI1H8 | |
Unconjugated | |
noggin, SYM1, symphalangism 1 (proximal), synostoses (multiple) syndrome 1, SYNS1 | |
Mouse | |
23.7 kDa | |
0.1 mL | |
Neuroscience, Stem Cell Markers, Stem Cell Signaling Pathway | |
9241 | |
Human, Mouse, Rat | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction