Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Noggin Antibody, PerCP, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB11040413PCP
Description
Noggin Polyclonal antibody specifically detects Noggin in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| Noggin | |
| Polyclonal | |
| Western Blot | |
| noggin, SYM1, symphalangism 1 (proximal), synostoses (multiple) syndrome 1, SYNS1 | |
| Synthetic peptide comprising residues 156-170 [PVLYAWNDLGSRFWP] of the human Noggin protein | |
| 0.1 mL | |
| Biologically Active Proteins, Neuroscience, Stem Cell Markers, Stem Cell Signaling Pathway | |
| 9241 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot | |
| PerCP | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction