Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Noggin Monoclonal Antibody (A11)
Mouse Monoclonal Antibody
$243.00 - $552.50
Specifications
Antigen | Noggin |
---|---|
Clone | A11 |
Concentration | 1 mg/mL |
Content And Storage | -20°C |
Applications | Immunohistochemistry (Paraffin), Western Blot |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIMA541654
|
Invitrogen™
MA541654 |
200 μL |
Each for $552.50
|
|
|||||
PIMA543714
|
Invitrogen™
MA543714 |
20 μL |
Each for $243.00
|
|
|||||
Description
Noggin Monoclonal Antibody for Western Blot, IHC (P)
Noggin is involved in numerous developmental processes, such as neural tube fusion and joint formation. The morphogenesis of organs is initiated by a downgrowth from a layer of epithelial stem cells. This process is achieved through the receipt of signals from 1) a WNT protein (WNT3A) to stabilize beta-catenin; and 2) Noggin, which is a bone morphogenetic protein inhibitor. Noggin mutations in unrelated families with proximal symphalangism (SYM1) and multiple synostoses syndrome (SYNS1) have been identified, which have multiple joint fusion as their principal defect.Specifications
Noggin | |
1 mg/mL | |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Mouse | |
RUO | |
PBS with 50% glycerol and 0.05% ProClin 300 | |
Q13253 | |
9241 | |
Recombinant protein Noggin. The antigen corresponds to amino acid range 28-232 of the target protein. | |
Antibody |
A11 | |
-20°C | |
Monoclonal | |
Liquid | |
IgG2b κ | |
Human | |
NOG | |
NOG; Noggin; Noggin precursor; noggin; LOW QUALITY PROTEIN: noggin; SYM1; symphalangism 1 (proximal); SYNS1; SYNS1A | |
NOG | |
Primary | |
Protein A/G |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title