Learn More
Noggin Mouse anti-Human, Clone: OTI1H8, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF500029

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Noggin is involved in numerous developmental processes, such as neural tube fusion and joint formation. The morphogenesis of organs is initiated by a downgrowth from a layer of epithelial stem cells. This process is achieved through the receipt of signals from 1) a WNT protein (WNT3A) to stabilize beta-catenin; and 2) Noggin, which is a bone morphogenetic protein inhibitor. Noggin mutations in unrelated families with proximal symphalangism (SYM1) and multiple synostoses syndrome (SYNS1) have been identified, which have multiple joint fusion as their principal defect.Specifications
Noggin | |
Monoclonal | |
Unconjugated | |
NOG | |
SYM1, SYNS1, SYNS1A | |
Mouse | |
Affinity Chromatography | |
RUO | |
9241 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
OTI1H8 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q13253 | |
NOG | |
Human recombit protein fragment corresponding to amino acids 28-232 of human noggin produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2b |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.