Learn More
Invitrogen™ Noggin Polyclonal Antibody

Description
Suggested positive control: antigen standard for NOG (transient overexpression lysate). Immunogen sequence: PVLYAWNDLG SRFWP.
Specifications
Specifications
| Antigen | Noggin |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene | NOG |
| Gene Accession No. | P97466, Q13253, Q62809 |
| Gene Alias | NOG; Noggin; Noggin precursor; noggin; LOW QUALITY PROTEIN: noggin; SYM1; symphalangism 1 (proximal); SYNS1; SYNS1A |
| Gene Symbols | NOG |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.