Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Noggin Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA534595
Description
Not suitable for use with Immunohistochemistry - Free Floating. This antibody is predicted to react with horse, chicken and Xenopus laevis based on sequence homology.
Noggin is involved in numerous developmental processes, such as neural tube fusion and joint formation. The morphogenesis of organs is initiated by a downgrowth from a layer of epithelial stem cells. This process is achieved through the receipt of signals from 1) a WNT protein (WNT3A) to stabilize beta-catenin; and 2) Noggin, which is a bone morphogenetic protein inhibitor. Noggin mutations in unrelated families with proximal symphalangism (SYM1) and multiple synostoses syndrome (SYNS1) have been identified, which have multiple joint fusion as their principal defect.
Specifications
| Noggin | |
| Polyclonal | |
| Unconjugated | |
| NOG | |
| NOG; Noggin; Noggin precursor; noggin; LOW QUALITY PROTEIN: noggin; SYM1; symphalangism 1 (proximal); SYNS1; SYNS1A | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 18121, 9241 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.9 mg/mL | |
| PBS with 0.02% sodium azide; pH 7.4 | |
| P97466, Q13253 | |
| NOG | |
| Synthetic peptide conjugated to KLH derived from within residues 1 - 100 of Human Noggin. | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction