Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Nogo-A Antibody, DyLight 650, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP300061C
Description
Nogo-A Polyclonal specifically detects Nogo-A in Human samples. It is validated for ELISA.Specifications
Nogo-A | |
Polyclonal | |
50 mM Sodium Borate | |
Produced in rabbits immunized with purified, recombinant Nogo-A (NP_065393.1; Met1-Val185). | |
0.1 mL | |
Primary | |
Human | |
Purified |
ELISA | |
DyLight 650 | |
Rabbit | |
Affinity purified | |
RUO | |
57142 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction