Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Nogo-A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA520367
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA520367 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA520367 Supplier Invitrogen™ Supplier No. PA520367
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Despite its predicted molecular weight, NogoA typically migrates at 180kDa in an SDS-PAGE. A suggested positive control is human brain tissue lysate. PA5-20367 can be used with blocking peptide PEP-0481.

NogoA is a member of a family of integral membrane proteins termed reticulons that are thought to be involved in numerous disorders including neurodegenerative diseases. Reticulon proteins are known to regulate many cellular processes and interact with multiple proteins and receptors such as BACE. NogoA was initially identified as a myelin-associated neurite outgrowth inhibitor. It is highly expressed in oligodendrocytes in the white matter of the CNS, blocking its activity with antibodies or other factors results in improved axon regrowth and functional recovery in experimental CNS lesion models. NogoA has also been suggested to play a role in neurodegenerative diseases such as Amyotrophic lateral sclerosis, in which case NogoA is found at elevated levels in postmortem muscular samples, and multiple sclerosis (MS), in which case autoantibodies to NogoA have been found in serum and cerebrospinal fluid in MS patients.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Nogo-A
Applications Immunohistochemistry, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene Rtn4
Gene Accession No. Q99P72, Q9JK11, Q9NQC3
Gene Alias 1110020G17Rik; AA407876; AA409940; AA960376; ASY; C130026I10Rik; Foocen; glut4 vesicle 20 kDa protein; GLUT4 vesicle 20kDa protein; Human NogoA; KIAA0886; mKIAA0886; mKIAA4153; My043; My043 protein; Nbla00271; Nbla10545; neurite growth inhibitor 220; neurite outgrowth inhibitor; Neuroendocrine-specific protein; Neuroendocrine-specific protein C homolog; NgA; NI220/250; NI-220/250; NI-250; Nogo; Nogo protein; Nogo-A; Nogo-B; NOGOC; Nogo-C; NSP; NSP-CL; r; rat N; rat NogoA; reticulon 4; reticulon 5; reticulon-4; Reticulon-5; RTN4; RTN4-A; RTN4-B1; RTN4-B2; RTN4-C; RTN-x; SP1507; Vp20
Gene Symbols Rtn4
Host Species Rabbit
Immunogen A 19 amino acid peptide from near the center of human NogoA.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57142, 68585, 83765
Target Species Human, Mouse, Rat
Content And Storage Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.