Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Nogo Receptor/NgR Antibody (SR2112), Novus Biologicals™

Rabbit Monoclonal Antibody
$309.50 - $423.00
Specifications
Antigen | Nogo Receptor/NgR |
---|---|
Clone | SR2112 |
Dilution | Western Blot 1:1000-1:5000, Immunoprecipitation 1:50 |
Applications | Western Blot, Immunoprecipitation |
Classification | Monoclonal |
Description
Nogo Receptor/NgR Monoclonal antibody specifically detects Nogo Receptor/NgR in Human, Mouse samples. It is validated for Western Blot, ImmunoprecipitationSpecifications
Nogo Receptor/NgR | |
Western Blot 1:1000-1:5000, Immunoprecipitation 1:50 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse | |
NgR, Nogo receptor, Nogo-66 receptor, NOGORUNQ330/PRO526, reticulon 4 receptor, reticulon-4 receptor | |
A synthesized peptide derived from human Nogo Receptor/NgR (Uniprot #: Q9BZR6) | |
Primary | |
Store at -20° C. Avoid freeze/thaw cycles. |
SR2112 | |
Western Blot, Immunoprecipitation | |
Unconjugated | |
Rabbit | |
Neurodegeneration, Neuroscience | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
65078 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title