Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Nogo Receptor/NgR Rabbit anti-Human, Mouse, Clone: 1R9A3, Novus Biologicals™

Rabbit Monoclonal Antibody
$205.50 - $492.50
Specifications
Antigen | Nogo Receptor/NgR |
---|---|
Clone | 1R9A3 |
Dilution | Western Blot 1:500 - 1:2000 |
Applications | Western Blot |
Classification | Monoclonal |
Description
Nogo Receptor/NgR Monoclonal antibody specifically detects Nogo Receptor/NgR in Human, Mouse samples. It is validated for Western BlotSpecifications
Nogo Receptor/NgR | |
Western Blot 1:500 - 1:2000 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse | |
NgR, Nogo receptor, Nogo-66 receptor, NOGORUNQ330/PRO526, reticulon 4 receptor, reticulon-4 receptor | |
A synthetic peptide corresponding to a sequence within amino acids 300-400 of human Nogo Receptor/NgR (Q9BZR6). RLAANDLQGCAVATGPYHPIWTGRATDEEPLGLPKCCQPDAADKASVLEPGRPASAGNALKGRVPPGDSPPGNGSGPRHINDSPFGTLPGSAEPPLTAVRP | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
1R9A3 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Neurodegeneration, Neuroscience | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
65078 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title