Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Nogo Receptor Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA577029
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
NOGO (RTN4R) is a receptor for RTN4, OMG, and MAG. Its signaling mediates activation of Rho and downstream reorganization of the actin cytoskeleton. NOGO also mediates axonal growth inhibition and may play a role in regulating axonal regeneration and plasticity in the adult CNS. In addition, NOGO acts with RTN4 and LINGO1 to regulate neuronal precursor cell motility during cortical development.
Specifications
Nogo Receptor | |
Polyclonal | |
Unconjugated | |
RTN4R | |
HGNC:1860; NgR; Ngr1; nogo 66 receptor; Nogo receptor; nogo receptor; Nogo-66 receptor; Nogo66 receptor; nogo-66 receptor; Nogo66 receptor-1; Nogor; OTTHUMP00000197866; reticulon 4 receptor; reticulon-4 receptor; Rtn4r; UNQ330/PRO526 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
113912, 65078, 65079 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.2 | |
Q99M75, Q99PI8, Q9BZR6 | |
RTN4R | |
Recombinant full length Human Nogo Receptor. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction