Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Nogo Receptor Polyclonal Antibody

Catalog No. PI720193
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI720193 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI720193 Supplier Invitrogen™ Supplier No. 720193

Rabbit Polyclonal Antibody

This antibody is predicted to react with Monkey, Rat, Mouse.

NOGO (RTN4R) is a receptor for RTN4, OMG, and MAG. Its signaling mediates activation of Rho and downstream reorganization of the actin cytoskeleton. NOGO also mediates axonal growth inhibition and may play a role in regulating axonal regeneration and plasticity in the adult CNS. In addition, NOGO acts with RTN4 and LINGO1 to regulate neuronal precursor cell motility during cortical development.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Nogo Receptor
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% BSA, 30% glycerol and 0.09% sodium azide; pH 7.4
Gene RTN4R
Gene Accession No. Q99M75, Q99PI8, Q9BZR6
Gene Alias HGNC:1860; NgR; Ngr1; nogo 66 receptor; Nogo receptor; nogo receptor; Nogo-66 receptor; Nogo66 receptor; nogo-66 receptor; Nogo66 receptor-1; Nogor; OTTHUMP00000197866; reticulon 4 receptor; reticulon-4 receptor; Rtn4r; UNQ330/PRO526
Gene Symbols RTN4R
Host Species Rabbit
Immunogen Peptides corresponding to human NOGO-66 Receptor (aa399-413, 317-334).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 113912, 65078, 65079
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.