Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nogo Receptor Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501980089
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-0089 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-0089 Supplier Bioss Supplier No. BS0129R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

NOGO (RTN4R) is a receptor for RTN4, OMG, and MAG. Its signaling mediates activation of Rho and downstream reorganization of the actin cytoskeleton. NOGO also mediates axonal growth inhibition and may play a role in regulating axonal regeneration and plasticity in the adult CNS. In addition, NOGO acts with RTN4 and LINGO1 to regulate neuronal precursor cell motility during cortical development.

Specifications

Antigen Nogo Receptor
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene RTN4R
Gene Accession No. Q9BZR6
Gene Alias HGNC:1860; NgR; Ngr1; Nogo receptor; nogo receptor; Nogo-66 receptor; Nogo66 receptor; Nogo-66 receptor; Nogo66 receptor-1; Nogor; OTTHUMP00000197866; reticulon 4 receptor; Reticulon-4 receptor; Rtn4r; UNQ330/PRO526
Gene Symbols RTN4R
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from mouse Nogo Receptor.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 65078
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.