Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Nogo Receptor Recombinant Rabbit Monoclonal Antibody (M5)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA547935
Description
Specificity: The antibody binds to human and rat NgR1 with a Kd of 2.9 nM for both species (bivalent binding).
NOGO (RTN4R) is a receptor for RTN4, OMG, and MAG. Its signaling mediates activation of Rho and downstream reorganization of the actin cytoskeleton. NOGO also mediates axonal growth inhibition and may play a role in regulating axonal regeneration and plasticity in the adult CNS. In addition, NOGO acts with RTN4 and LINGO1 to regulate neuronal precursor cell motility during cortical development.
Specifications
Nogo Receptor | |
Recombinant Monoclonal | |
1 mg/mL | |
PBS with 0.02% ProClin 300 | |
Q99M75, Q99PI8, Q9BZR6 | |
RTN4R | |
(His)6-tagged fragment of rat NgR1 (amino acids 278-473). | |
200 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG κ |
ELISA, Flow Cytometry, Immunoprecipitation, Western Blot | |
M5 | |
Unconjugated | |
RTN4R | |
HGNC:1860; NgR; Ngr1; nogo 66 receptor; Nogo receptor; nogo receptor; Nogo-66 receptor; Nogo66 receptor; nogo-66 receptor; Nogo66 receptor-1; Nogor; OTTHUMP00000197866; reticulon 4 receptor; reticulon-4 receptor; Rtn4r; UNQ330/PRO526 | |
Rabbit | |
Protein A | |
RUO | |
113912, 65078, 65079 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction