Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NOL1 Antibody, Alexa Fluor™ 750, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP118907AF750
Description
NOL1 Polyclonal antibody specifically detects NOL1 in Human samples. It is validated for Western BlotSpecifications
NOL1 | |
Polyclonal | |
Western Blot | |
EC 2.1.1, EC 2.1.1.-, MGC149287, MGC149288, NOL1/NOP2/Sun domain family, member 1, NOL1MGC117384, NOP120, NOP2 nucleolar protein homolog (yeast), NOP2/Sun domain family, member 1, NSUN1, Nucleolar protein 1, nucleolar protein 1 (120kD), nucleolar protein 1, 120kDa, Nucleolar protein 2 homolog, nucleolar protein 2 homolog (yeast), p120, Proliferating-cell nucleolar antigen p120, Proliferation-associated nucleolar protein p120, putative ribosomal RNA methyltransferase NOP2 | |
The immunogen recognized by this antibody maps to a region between residue 762 and 812 of human nucleolar protein 1 using the numbering given in entry AAH00656.1 (GeneID 4839). | |
0.1 mL | |
Core ESC Like Genes, Stem Cell Markers | |
4839 | |
Store at 4°C in the dark. | |
IgG |
Western Blot | |
Alexa Fluor 750 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction