Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NOL1 Rabbit anti-Human, Polyclonal, Bioss

Rabbit Polyclonal Antibody
Supplier: Bioss BS9710R

Description
NOP2 is a member of the unc-84 homolog family and encodes a nuclear nuclear envelope protein with an Unc84 (SUN) domain. The protein is involved in nuclear anchorage and migration. Alternatively spliced transcript variants have been described.Specifications
NOL1 | |
Polyclonal | |
Unconjugated | |
Nop2 | |
120kDa; 25S rRNA (cytosine(2870)-C(5))-methyltransferase; A530002O17; BOS_5976; cb164; im:6893299; MGC117384; MGC149287; MGC149288; N2428; NOL1; NOL1/NOP2/Sun domain family, member 1; NOP120; NOP2; NOP2 nucleolar protein; NOP2 nucleolar protein homolog; NOP2 nucleolar protein homolog (yeast); nop2p; NSUN1; Nucleolar protein 1; nucleolar protein 1, 120 kDa; nucleolar protein 1, 120kDa; Nucleolar protein 2; Nucleolar protein 2 homolog; p120; Probable 28S rRNA (cytosine(4447)-C(5))-methyltransferase; probable 28S rRNA (cytosine-C(5))-methyltransferase; Proliferating-cell nucleolar antigen p120; Proliferation-associated nucleolar protein p120; putative ribosomal RNA methyltransferase NOP2; rRNA (cytosine-C5-)-methyltransferase NOP2; sb:cb164; si:dkey-91i17.1; Unknown (protein for IMAGE:6909257); YNA1; YNL061W; YNL2428W | |
Rabbit | |
Protein A | |
RUO | |
4839 | |
-20°C | |
Liquid |
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
1 μg/mL | |
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH | |
P46087 | |
Nop2 | |
KLH conjugated synthetic peptide derived from human NOL1. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction