Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NOL11 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA554549
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA554549 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA554549 Supplier Invitrogen™ Supplier No. PA554549
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: FTLSSVVLSA GPEGLLGVEQ SDKTDQFLVT DSGRTVILYK VSDQKPLGSW SVKQGQIITC PAVCNFQTGE YVVVHDNKVL RIWNNEDVNL Highest antigen sequence identity to the following orthologs: Mouse - 86%, Rat - 87%.

NOL11 is a ribosome biogenesis factor. It may be required for both optimal rDNA transcription and small subunit (SSU) pre-rRNA processing at sites A', A0, 1 and 2b.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NOL11
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene NOL11
Gene Accession No. Q9H8H0
Gene Alias 1500002M01Rik; AU015220; AU019874; L14; Nol11; Nucleolar protein 11
Gene Symbols NOL11
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human NOL11. Recombinant protein control fragment (Product #RP-93253).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25926
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.