Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NOL4 Monoclonal Antibody (2A10)
GREENER_CHOICE

Catalog No. PIMA524429
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA524429 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA524429 Supplier Invitrogen™ Supplier No. MA524429
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Peptide Sequence: PGSQDVLYIN GNGTYSYHSY RGLGGGLLNL NDASSSGPTD LSMKRQLATS SGSSSSSNSR PQLSPTEINA VRQLVAGYRE SAAFLLRSAD ELENLILQQN.

NOL4 gene ontology (GO) annotation includes RNA binding and nucleolus.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NOL4
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 2A10
Concentration 0.48 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene Nol4
Gene Accession No. O94818
Gene Alias 1700013J13Rik; 4930568N03Rik; cancer/testis antigen 125; CT125; Gm1262; HRIHFB2255; Nol4; NOLP; nucleolar localized protein; nucleolar protein 4; Nucleolar-localized protein
Gene Symbols Nol4
Host Species Mouse
Immunogen NOL4 (NP_003778, 425 a.a. ~ 524 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8715
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.