Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NOL4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5103986
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5103986 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5103986 Supplier Invitrogen™ Supplier No. PA5103986
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total NOL4.

NOL4 gene ontology (GO) annotation includes RNA binding and nucleolus.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NOL4
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene Nol4
Gene Accession No. O94818, P60954
Gene Alias 1700013J13Rik; 4930568N03Rik; cancer/testis antigen 125; CT125; Gm1262; HRIHFB2255; Nol4; NOLP; nucleolar localized protein; nucleolar protein 4; Nucleolar-localized protein
Gene Symbols Nol4
Host Species Rabbit
Immunogen A synthesized peptide derived from human NOL4(Accession O94818), corresponding to amino acid residues Y190-S240.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 307553, 319211, 8715
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.