Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ NOL6 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA563202
Description
Immunogen sequence: GPSSLMPVLG YDPPQLYLTQ LREAFGDLAL FFYDQHGGEV IGVLWKPTSF QPQPFKASST KGRMVMSRGG ELVMVPNVEA ILEDFAVLGE GLVQTVEARS ERWTV Highest antigen sequence identity to the following orthologs: Mouse - 91%, Rat - 93%.
The nucleolus is a dense subnuclear membraneless organelle that assembles around clusters of rRNA genes and functions in ribosome biogenesis. This gene encodes a nucleolar RNA-associated protein that is highly conserved between species. RNase treatment of permeabilized cells indicates that the nucleolar localization is RNA dependent. Further studies suggest that the protein is associated with ribosome biogenesis through an interaction with pre-rRNA primary transcripts. Alternative splicing has been observed at this locus and two splice variants encoding distinct isoforms have been identified.
Specifications
NOL6 | |
Polyclonal | |
Unconjugated | |
NOL6 | |
AA410091; bA311H10.1; LOW QUALITY PROTEIN: nucleolar protein 6; Nol6; Nrap; nucleolar protein 6; nucleolar protein 6 (RNA-associated); nucleolar protein family 6; nucleolar protein family 6 (RNA-associated); nucleolar RNA-associated protein; UTP22 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
65083 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9H6R4 | |
NOL6 | |
Recombinant protein corresponding to Human NOL6. Recombinant protein control fragment (Product #RP-93046). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction