Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NOL6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA584476
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA584476 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA584476 Supplier Invitrogen™ Supplier No. PA584476
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PQVGFLRFLF LVSTFDWKNN PLFVNLNNEL TVEEQVEIRS GFLAARAQLP VMVIVTPQDR KNSVWTQDGP SAQILQQLVV LAAEALPML Highest antigen sequence indentity to the following orthologs: Rat - 88%, Mouse - 87%.

The nucleolus is a dense subnuclear membraneless organelle that assembles around clusters of rRNA genes and functions in ribosome biogenesis. This gene encodes a nucleolar RNA-associated protein that is highly conserved between species. RNase treatment of permeabilized cells indicates that the nucleolar localization is RNA dependent. Further studies suggest that the protein is associated with ribosome biogenesis through an interaction with pre-rRNA primary transcripts. Alternative splicing has been observed at this locus and two splice variants encoding distinct isoforms have been identified.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NOL6
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene NOL6
Gene Accession No. Q9H6R4
Gene Alias AA410091; bA311H10.1; LOW QUALITY PROTEIN: nucleolar protein 6; Nol6; Nrap; nucleolar protein 6; nucleolar protein 6 (RNA-associated); nucleolar protein family 6; nucleolar protein family 6 (RNA-associated); nucleolar RNA-associated protein; UTP22
Gene Symbols NOL6
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human NOL6. Recombinant protein control fragment (Product #RP-105164).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 65083
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.