Learn More
Invitrogen™ NOL7 Polyclonal Antibody

Description
Immunogen sequence: EPLEEDEEGD DEFDDEAPEE LTFASAQAEA REEERRVRET VRRDKTLLKE KRKRREELFI EQKKRKL Highest antigen sequence identity to the following orthologs: Mouse - 82%, Rat - 85%.
Specifications
Specifications
| Antigen | NOL7 |
| Applications | Immunohistochemistry (Paraffin), Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.3 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | NOL7 |
| Gene Accession No. | Q9UMY1 |
| Gene Alias | 2210008F15Rik; 5730556I21Rik; C6orf90; dJ223E5.2; MNCb-1192; NOL7; Nol7-ps1; NOP27; nucleolar protein 7; nucleolar protein 7, 27kDa; nucleolar protein 7, pseudogene 1; nucleolar protein of 27 kDa; polyglutamine binding protein 3; PQBP3; RARG-1; retinoic acid repressible protein; RGD1565934 |
| Gene Symbols | NOL7 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.