Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NOL8 Antibody, Alexa Fluor™ 594, Novus Biologicals™
SDP

Catalog No. NB260964 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB260964 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB260964 Supplier Novus Biologicals Supplier No. NBP146849AF594
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

NOL8 Polyclonal antibody specifically detects NOL8 in Human samples. It is validated for Western Blot, Immunohistochemistry

Specifications

Antigen NOL8
Applications Western Blot, Immunohistochemistry
Classification Polyclonal
Conjugate Alexa Fluor 594
Dilution Western Blot, Immunohistochemistry
Formulation 50mM Sodium Borate
Gene Alias NOP132, nucleolar protein 8
Host Species Rabbit
Immunogen The immunogen for this product maps to a region between residue 200 and 250 of human nucleolar protein Nop132 using the numbering given in entry NP_060418.4 (GeneID 55035).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55035
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.