Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NOL8 Antibody, DyLight 680, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP146850FR
Description
NOL8 Polyclonal antibody specifically detects NOL8 in Human samples. It is validated for Western Blot, ImmunohistochemistrySpecifications
NOL8 | |
Polyclonal | |
Western Blot, Immunohistochemistry | |
NOP132, nucleolar protein 8 | |
The immunogen for this product maps to a region between residue 1117 and 1167 of human nucleolar protein Nop132 using the numbering given in entry NP_060418.4 (GeneID 55035). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot, Immunohistochemistry | |
DyLight 680 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
55035 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction